Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191056.3 | NP_001177985.1 | 9 | 1339 | ||
NM_001191057.3 | NP_001177986.1 | 9 | 1339 | |||
NM_001191058.3 | NP_001177987.2 | 10 | 1719 | |||
NM_001191059.3 | NP_001177988.1 | 10 | 1330 | |||
NM_001322055.1 | NP_001308984.1 | 10 | 1565 | |||
NM_001322056.1 | NP_001308985.1 | 10 | 1330 | |||
NM_001322057.1 | NP_001308986.1 | 10 | 1565 | |||
NM_001322058.1 | NP_001308987.1 | 10 | 1719 | |||
NM_001322059.1 | NP_001308988.1 | 10 | 1368 | |||
NM_005020.4 | NP_005011.1 | 10 | 1103 | |||
XM_017012264.1 | XP_016867753.1 | 10 | 1170 | |||
XM_017012265.1 | XP_016867754.1 | 10 | 1021 | |||
XM_017012266.1 | XP_016867755.1 | 10 | 1170 | |||
XM_017012267.1 | XP_016867756.1 | 10 | 1075 | |||
GenBank mRNA | AK056170.1 | - | 9 | 1326 | ||
AK091734.1 | - | 10 | 1644 | |||
AK094180.1 | - | 10 | 1262 | |||
BC022479.1 | - | 9 | 1340 | 4795813 | ||
BC022525.1 | - | 9 | 1340 | 4795158 | ||
U40371.1 | - | 10 | 1046 | |||
U40372.1 | - | 5 | 436 |