Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143998
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
31
5545
NM_005358.5
NP_005349.3
30
5589
NM_015842.2
NP_056667.2
27
5127
XM_011535075.2
XP_011533377.1
32
5433
XM_011535078.1
XP_011533380.1
31
5391
XM_011535080.2
XP_011533382.1
32
5352
XM_011535082.1
XP_011533384.1
31
5310
XM_011535086.1
XP_011533388.1
29
4997
XM_011535094.1
XP_011533396.1
25
4014
XM_011535095.1
XP_011533397.1
28
4185
XM_017020574.1
XP_016876063.1
31
6745
XM_017020575.1
XP_016876064.1
30
6703
XM_017020576.1
XP_016876065.1
30
6688
XM_017020577.1
XP_016876066.1
31
6664
XM_017020578.1
XP_016876067.1
29
6646
XM_017020579.1
XP_016876068.1
30
6622
XM_017020580.1
XP_016876069.1
30
6541
XM_017020581.1
XP_016876070.1
30
6667
XM_017020582.1
XP_016876071.1
30
6460
XM_017020583.1
XP_016876072.1
31
5148
XM_017020584.1
XP_016876073.1
29
6463
XM_017020585.1
XP_016876074.1
29
6259
XM_017020586.1
XP_016876075.1
29
6178
XM_017020587.1
XP_016876076.1
28
6136
XM_017020588.1
XP_016876077.1
28
4796
XM_017020589.1
XP_016876078.1
28
4919
XM_017020590.1
XP_016876079.1
28
4715
XM_017020591.1
XP_016876080.1
30
6828
XM_017020592.1
XP_016876081.1
30
6711
XM_017020593.1
XP_016876082.1
29
6624
XM_017020594.1
XP_016876083.1
29
6507
XM_017020595.1
XP_016876084.1
29
5189
XM_017020596.1
XP_016876085.1
28
5539
XM_017020597.1
XP_016876086.1
27
5509
XM_017020598.1
XP_016876087.1
28
5080
XM_017020599.1
XP_016876088.1
28
6342
XM_017020600.1
XP_016876089.1
29
4146
XM_017020601.1
XP_016876090.1
28
4907
XM_017020602.1
XP_016876091.1
28
4790
XM_017020603.1
XP_016876092.1
27
4798
XM_017020604.1
XP_016876093.1
27
5622
GenBank mRNA
AB020665.2
-
27
5057
AF144237.1
-
19
3049
AF330045.1
-
30
5588
AK074265.1
-
15
2041
U90654.1
-
8
1305
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0