Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005372.1 | NP_005363.1 | 1 | 340 | ||
GenBank mRNA | BC069569.1 | - | 1 | 340 | 7262233 | |
BC069590.1 | - | 1 | 340 | |||
BC106737.1 | - | 1 | 340 | 40016104 | ||
BC106738.1 | - | 1 | 340 | 40016106 |