Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163257.1 | NP_001156729.1 | 29 | 5145 | ||
NM_005393.2 | NP_005384.2 | 28 | 4979 | |||
GenBank mRNA | AB033032.1 | - | 28 | 4919 | ||
AF149019.1 | - | 28 | 4919 | |||
AK292535.1 | - | 28 | 4979 | |||
AK295762.1 | - | 28 | 4999 | |||
AK295894.1 | - | 27 | 3863 | |||
AK307894.1 | - | 29 | 5122 |