Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005558.3 | NP_005549.2 | 10 | 2765 | ||
GenBank mRNA | BC009742.2 | - | 10 | 2765 | 3842503 |