Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005565.3 | NP_005556.1 | 9, 10 | 887 | ||
GenBank mRNA | AK292890.1 | - | 9, 10 | 848 | ||
AK296098.1 | - | 1, 2 | 244 | |||
AK300000.1 | - | 7, 8 | 731 | |||
AK310369.1 | - | 6, 7 | 498 | |||
BC016618.1 | - | 9, 10 | 824 | 4476274 | ||
BT007273.1 | - | 9, 10 | 680 | |||
FN432830.1 | - | 10, 11 | 760 | |||
U20158.1 | - | 9, 10 | 935 |