Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005565.3 | NP_005556.1 | 13 | 1046 | ||
GenBank mRNA | AK292890.1 | - | 13 | 1007 | ||
AK296098.1 | - | 5 | 403 | |||
AK300000.1 | - | 11 | 890 | |||
AK310369.1 | - | 10 | 657 | |||
BC016618.1 | - | 13 | 983 | 4476274 | ||
BT007273.1 | - | 13 | 839 | |||
FN432830.1 | - | 14 | 919 | |||
U20158.1 | - | 13 | 1094 |