Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005582.2 | NP_005573.2 | 3 | 759 | ||
XM_005248504.4 | XP_005248561.1 | 4 | 768 | |||
GenBank mRNA | AK313177.1 | - | 3 | 769 | ||
AL544501.3 | - | 3 | 759 | |||
BC109069.1 | - | 3 | 717 | 40034168 | ||
BC109070.1 | - | 3 | 717 | 40034172 | ||
BP344300.1 | - | 2 | 421 | |||
D83597.1 | - | 3 | 753 |