Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005584.4 | NP_005575.1 | 1 | 1845 | ||
GenBank mRNA | AB073388.1 | - | 1 | 1835 | ||
AB593021.1 | - | 1 | 1766 | |||
AB593022.1 | - | 1 | 1836 | |||
BC028170.1 | - | 1 | 1766 | 5244028 |