Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166280.1 | NP_001159752.1 | 5 | 663 | ||
NM_001166281.1 | NP_001159753.1 | 5 | 663 | |||
NM_005592.3 | NP_005583.1 | 5 | 663 | |||
XM_005251994.3 | XP_005252051.1 | 5 | 1245 | |||
XM_005251995.3 | XP_005252052.1 | 5 | 1244 | |||
XM_005251996.3 | XP_005252053.1 | 5 | 1243 | |||
XM_017014734.1 | XP_016870223.1 | 5 | 1245 | |||
GenBank mRNA | AF006464.1 | - | 5 | 575 | ||
BC109098.1 | - | 5 | 575 | 40034772 | ||
BC109099.1 | - | 5 | 575 | 40034773 |