Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005598.3 | NP_005589.1 | 2 | 1627 | ||
GenBank mRNA | BC013789.1 | - | 2 | 1614 | 3866861 | |
M96739.1 | - | 2 | 1630 |