Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190821.1 | NP_001177750.1 | 4 | 1605 | ||
NM_001190822.1 | NP_001177751.1 | 4 | 813 | |||
NM_001190823.1 | NP_001177752.1 | 2 | 906 | |||
NM_005904.3 | NP_005895.1 | 4 | 1608 | |||
GenBank mRNA | AF010193.1 | - | 4 | 1616 | ||
AK131394.1 | - | 2 | 848 | |||
AK301535.1 | - | 4 | 813 | |||
BC074818.2 | - | 4 | 1355 | 30915321 | ||
BC074819.2 | - | 4 | 1355 | 30915535 |