Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005907.3 | NP_005898.2 | 4 | 1170 | ||
XM_005266986.4 | XP_005267043.1 | 4 | 1026 | |||
XM_011535833.2 | XP_011534135.1 | 3 | 222 | |||
XM_011535834.2 | XP_011534136.1 | 4 | 1038 | |||
GenBank mRNA | AK025599.1 | - | 4 | 1165 | ||
AK307383.1 | - | 4 | 1165 | |||
BC065827.1 | - | 5 | 1209 | 30390733 |