Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 13 | 2652 | ||
NM_005916.4 | NP_005907.3 | 14 | 2577 | |||
NM_182776.2 | NP_877577.1 | 13 | 2543 | |||
XM_005250348.3 | XP_005250405.1 | 14 | 2231 | |||
XM_017012217.1 | XP_016867706.1 | 14 | 2108 | |||
GenBank mRNA | AK055379.1 | - | 13 | 2520 | ||
AK096959.1 | - | 15 | 1941 | |||
AK226175.1 | - | 13 | 2082 | |||
AK293172.1 | - | 14 | 1979 | |||
AK314575.1 | - | 14 | 2044 | |||
AY007130.1 | - | 6 | 874 | |||
BC013375.2 | - | 14 | 2013 | 3946342 | ||
D55716.1 | - | 14 | 2047 | |||
D86748.1 | - | 14 | 2017 | |||
KU178172.1 | - | 14 | 1931 | |||
KU178173.1 | - | 13 | 2006 | |||
X74796.1 | - | 11 | 1627 |