Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291958.1 | NP_001278887.1 | 2 | 535 | ||
NM_001301072.1 | NP_001288001.1 | 2 | 535 | |||
NM_005922.3 | NP_005913.2 | 2 | 535 | |||
NM_006724.3 | NP_006715.2 | 2 | 535 | |||
NR_120425.1 | 2 | 535 | ||||
XM_005266989.2 | XP_005267046.1 | 2 | 739 | |||
XM_017010869.1 | XP_016866358.1 | 3 | 959 | |||
GenBank mRNA | AF002715.1 | - | 2 | 472 | ||
AK307866.1 | - | 2 | 404 | |||
BC136276.1 | - | 2 | 422 | 9020263 | ||
BC143735.1 | - | 2 | 419 | 9052249 | ||
BC143743.1 | - | 2 | 418 | |||
BC143750.1 | - | 2 | 418 | |||
BC146770.1 | - | 2 | 535 | 9007216 | ||
D86968.2 | - | 2 | 535 |