Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005923.3 | NP_005914.1 | 26, 27 | 4124 | ||
XM_017010870.1 | XP_016866359.1 | 27, 28 | 4450 | |||
XM_017010871.1 | XP_016866360.1 | 27, 28 | 4447 | |||
XM_017010872.1 | XP_016866361.1 | 27, 28 | 4449 | |||
XM_017010873.1 | XP_016866362.1 | 27, 28 | 4445 | |||
XM_017010874.1 | XP_016866363.1 | 26, 27 | 4342 | |||
XM_017010875.1 | XP_016866364.1 | 26, 27 | 4338 | |||
XM_017010876.1 | XP_016866365.1 | 25, 26 | 3458 | |||
XM_017010877.1 | XP_016866366.1 | 25, 26 | 3695 | |||
GenBank mRNA | AB209093.1 | - | 26, 27 | 4231 | ||
AK293953.1 | - | 26, 27 | 3387 | |||
AK294507.1 | - | 16, 17 | 2106 | |||
BC054503.2 | - | 26, 27 | 4071 | 6143181 | ||
BC088829.1 | - | 26, 27 | 4125 | 6007002 | ||
D84476.2 | - | 26, 27 | 4030 | |||
U67156.1 | - | 26, 27 | 4123 |