Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135037.1 | NP_001128509.1 | 2 | 1271 | ||
NM_001242336.1 | NP_001229265.1 | 3 | 2029 | |||
NM_005927.4 | NP_005918.1 | 3 | 1732 | |||
GenBank mRNA | AK000358.1 | - | 2 | 1693 | ||
BC026244.1 | - | 3 | 1989 | 4697443 |