Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001298.1 | 1 | 119 | |||
GenBank mRNA | AF005487.1 | - | 1 | 119 | ||
L76565.1 | - | 1 | 119 | |||
U33547.1 | - | 1 | 139 |
Gene Symbol: MICB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289160.1 | NP_001276089.1 | 6 | 1552 | ||
NM_001289161.1 | NP_001276090.1 | 6 | 1522 | |||
NM_005931.4 | NP_005922.2 | 6 | 1651 | |||
GenBank mRNA | AK303655.1 | - | 5 | 1784 | ||
BC044218.1 | - | 6 | 1484 | 6025253 | ||
CN389975.1 | - | 1 | 501 | |||
X91625.1 | - | 6 | 1510 |