Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017973.1 | NP_001017973.1 | 4, 5 | 903 | ||
NM_001017974.1 | NP_001017974.1 | 4, 5 | 431 | |||
NM_001142598.1 | NP_001136070.1 | 5, 6 | 567 | |||
NM_001142599.1 | NP_001136071.1 | 5, 6 | 567 | |||
NM_004199.2 | NP_004190.1 | 4, 5 | 903 | |||
XM_005272116.4 | XP_005272173.1 | 4, 5 | 431 | |||
XM_005272117.4 | XP_005272174.1 | 5, 6 | 589 | |||
XM_005272118.4 | XP_005272175.1 | 5, 6 | 718 | |||
XM_005272119.4 | XP_005272176.1 | 5, 6 | 908 | |||
XM_005272120.4 | XP_005272177.1 | 5, 6 | 688 | |||
XM_006714728.3 | XP_006714791.1 | 5, 6 | 965 | |||
XM_006714729.3 | XP_006714792.1 | 5, 6 | 589 | |||
XM_006714730.3 | XP_006714793.1 | 5, 6 | 965 | |||
XM_011543705.2 | XP_011542007.1 | 3, 4 | 357 | |||
XM_017010011.1 | XP_016865500.1 | 5, 6 | 688 | |||
XM_017010012.1 | XP_016865501.1 | 4, 5 | 431 | |||
GenBank mRNA | AU117989.1 | - | 4, 5 | 431 | ||
AY358970.1 | - | 4, 5 | 505 | |||
BC017062.1 | - | 4, 5 | 532 | 3847723 | ||
BC035813.1 | - | 4, 5 | 400 | 5753499 | ||
BI758477.1 | - | 5, 6 | 518 | |||
BX647924.1 | - | 4, 5 | 423 | |||
U90441.1 | - | 4, 5 | 526 |