Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004196.3 | NP_001004196.2 | 7 | 1283 | ||
NM_001318826.1 | NP_001305755.1 | 6 | 1203 | |||
NM_001318828.1 | NP_001305757.1 | 5 | 1126 | |||
NM_001318830.1 | NP_001305759.1 | 5 | 1121 | |||
NM_005944.6 | NP_005935.4 | 6 | 1208 | |||
XM_017006454.1 | XP_016861943.1 | 7 | 1773 | |||
XM_017006455.1 | XP_016861944.1 | 6 | 1408 | |||
XM_017006456.1 | XP_016861945.1 | 7 | 1134 | |||
XM_017006458.1 | XP_016861947.1 | 7 | 1774 | |||
XM_017006459.1 | XP_016861948.1 | 6 | 1393 | |||
GenBank mRNA | AF063591.1 | - | 6 | 999 | ||
AK074972.1 | - | 6 | 973 | |||
AK129797.1 | - | 5 | 890 | |||
AK293399.1 | - | 5 | 895 | |||
AK297194.1 | - | 7 | 1074 | |||
AL832529.1 | - | 6 | 993 | |||
BC022522.1 | - | 6 | 1001 | 4794050 | ||
BC031103.1 | - | 6 | 977 | 5301702 |