Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005969.3 | NP_005960.1 | 10 | 953 | ||
GenBank mRNA | AB500094.1 | - | 8 | 812 | ||
AK299515.1 | - | 10 | 876 | |||
AK312599.1 | - | 10 | 961 | |||
AK316242.1 | - | 10 | 870 | |||
AK316324.1 | - | 10 | 891 | |||
AK316548.1 | - | 11 | 1010 | |||
BC022090.2 | - | 10 | 863 | 3029455 | ||
U77456.1 | - | 10 | 961 |