Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001315529.1 | NP_001302458.1 | 3 | 781 | ||
NM_006031.5 | NP_006022.3 | 3 | 619 | |||
XM_005261124.4 | XP_005261181.1 | 3 | 638 | |||
XM_011529594.2 | XP_011527896.1 | 3 | 638 | |||
XM_017028362.1 | XP_016883851.1 | 3 | 638 | |||
XM_017028363.1 | XP_016883852.1 | 3 | 807 | |||
GenBank mRNA | AB007862.3 | - | 3 | 782 | ||
AF515282.1 | - | 3 | 512 | |||
AK024009.1 | - | 4 | 561 | |||
BC035913.1 | - | 4 | 561 | |||
U52962.1 | - | 3 | 564 |