Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190796.2 | NP_001177725.1 | 2 | 490 | ||
NM_001291999.1 | NP_001278928.1 | 3 | 760 | |||
NM_006153.5 | NP_006144.1 | 3 | 726 | |||
GenBank mRNA | AK301460.1 | - | 2 | 448 | ||
AK312692.1 | - | 3 | 670 | |||
BC006403.2 | - | 3 | 703 | 4304621 | ||
BP339594.1 | - | 2 | 491 | |||
KU178218.1 | - | 2 | 596 | |||
X17576.1 | - | 3 | 697 |