Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEURL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004210.4 | NP_004201.3 | 3 | 910 | ||
XM_005270269.3 | XP_005270326.1 | 3 | 579 | |||
XM_005270270.3 | XP_005270327.1 | 3 | 556 | |||
XM_011540331.2 | XP_011538633.1 | 4 | 1112 | |||
XM_011540332.2 | XP_011538634.1 | 4 | 759 | |||
XM_011540333.2 | XP_011538635.1 | 3 | 645 | |||
XM_011540335.2 | XP_011538637.1 | 4 | 1112 | |||
XM_017016909.1 | XP_016872398.1 | 3 | 539 | |||
GenBank mRNA | AF029729.1 | - | 3 | 561 | ||
BC026336.1 | - | 3 | 597 | 4812302 | ||
U87864.1 | - | 3 | 910 |