Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006204.3 | NP_006195.3 | 3 | 820 | ||
GenBank mRNA | U31973.1 | - | 3 | 820 |