Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006205.2 | NP_006196.1 | 4 | 454 | ||
XM_017019431.1 | XP_016874920.1 | 3 | 697 | |||
GenBank mRNA | BC069774.1 | - | 3 | 391 | 7262475 | |
BC069800.1 | - | 3 | 391 | 7262487 | ||
D45399.1 | - | 4 | 432 | |||
EL946662.1 | - | 4 | 458 |