Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006209.4 | NP_006200.3 | 11, 12 | 1066 | ||
XM_006716584.1 | XP_006716647.1 | 11, 12 | 1044 | |||
XM_006716585.1 | XP_006716648.1 | 11, 12 | 1044 | |||
XM_006716587.1 | XP_006716650.1 | 11, 12 | 1044 | |||
GenBank mRNA | L35594.1 | - | 11, 12 | 1029 |