Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170747.1 | NP_001164218.1 | 2 | 215 | ||
NM_006223.3 | NP_006214.2 | 2 | 215 | |||
GenBank mRNA | AB009690.1 | - | 2 | 136 | ||
AF143096.1 | - | 2 | 129 | |||
AK127605.1 | - | 2 | 126 | |||
AU098526.1 | - | 2 | 215 | |||
AV709926.2 | - | 2 | 164 | |||
BC005234.1 | - | 2 | 134 | |||
BC070288.1 | - | 2 | 134 | |||
BC093700.1 | - | 2 | 212 | 7939545 | ||
BC104653.1 | - | 2 | 135 | |||
BC111394.1 | - | 2 | 138 | |||
BC112281.1 | - | 2 | 212 | 8327749 |
Gene Symbol: PIN4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003571.1 | 1 | 2032 | |||
GenBank mRNA | AK055299.1 | - | 1 | 2032 |