Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198909.1 | NP_001185838.1 | 3 | 722 | ||
NM_004231.3 | NP_004222.2 | 2 | 638 | |||
GenBank mRNA | BC107854.1 | - | 2 | 615 | 5171984 | |
BG724326.1 | - | 2 | 624 | |||
BU956696.1 | - | 3 | 621 | |||
D49400.1 | - | 2 | 586 |