Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012394.3 | NP_036526.2 | 2, 3 | 217 | ||
XM_011509624.2 | XP_011507926.1 | 3, 4 | 230 | |||
GenBank mRNA | AF117237.1 | - | 2, 3 | 173 | ||
AF151065.1 | - | 2, 3 | 157 | |||
AF165883.1 | - | 2, 3 | 190 | |||
AK024531.1 | - | 2, 3 | 197 | |||
AK315736.1 | - | 2, 3 | 191 | |||
BC012464.1 | - | 2, 3 | 186 | 4538003 | ||
BC047042.1 | - | 2, 3 | 183 | 5541226 | ||
BI597083.1 | - | 2, 3 | 250 |