Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145354.1 | NP_001138826.1 | 19 | 8112 | ||
NM_001321316.1 | NP_001308245.1 | 18 | 8043 | |||
NM_013255.4 | NP_037387.2 | 18 | 8040 | |||
XM_006715993.2 | XP_006716056.1 | 18 | 7964 | |||
GenBank mRNA | AK074367.1 | - | 1 | 193 | ||
AL833458.1 | - | 1 | 146 | |||
AW505004.1 | - | 1 | 370 |