Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014057.4 | NP_054776.1 | 7 | 1055 | ||
NM_024416.4 | NP_077727.3 | 7 | 974 | |||
NM_033014.3 | NP_148935.1 | 7 | 1337 | |||
GenBank mRNA | AF086912.1 | - | 7 | 902 | ||
AF088023.1 | - | 4 | 393 | |||
AF100758.1 | - | 7 | 902 | |||
AF202167.1 | - | 6 | 827 | |||
AK129845.1 | - | 6 | 859 | |||
AK289628.1 | - | 7 | 1052 | |||
AK295433.1 | - | 7 | 974 | |||
AL110267.1 | - | 6 | 844 | |||
AL133114.1 | - | 3 | 320 | |||
BC037273.2 | - | 6 | 843 | 5265880 | ||
BC095443.1 | - | 6 | 859 | 30405707 | ||
CR533516.1 | - | 6 | 776 |