Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBD1 SIRNA Assays ›
Gene Symbol:
MBD1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
CXXC3, PCM1, RFT
Chromosome Location:
Chr.18: 50266882 - 50281774 on Build GRCh38
UniGene ID:
Hs.405610
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144323
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204136.1
NP_001191065.1
12
1810
NM_001204137.1
NP_001191066.1
13
1885
NM_001204138.1
NP_001191067.1
13
1882
NM_001204139.1
NP_001191068.1
12
1810
NM_001204140.1
NP_001191069.1
12
1717
NM_001204141.1
NP_001191070.1
11
1660
NM_001204142.1
NP_001191071.1
12
1810
NM_001204143.1
NP_001191072.1
11
1423
NM_001204151.2
NP_001191080.1
11
1741
NM_001323942.1
NP_001310871.1
13
1885
NM_001323947.1
NP_001310876.1
13
1885
NM_001323949.1
NP_001310878.1
13
1877
NM_001323950.1
NP_001310879.1
12
1807
NM_001323951.1
NP_001310880.1
12
1810
NM_001323952.1
NP_001310881.1
11
1567
NM_001323953.1
NP_001310882.1
11
1748
NM_001323954.1
NP_001310883.1
10
1573
NM_002384.2
NP_002375.1
11
1642
NM_015844.2
NP_056669.2
11
1642
NM_015845.3
NP_056670.2
11
1741
NM_015846.3
NP_056671.2
12
1810
NM_015847.3
NP_056723.2
11
1663
XM_005258271.2
XP_005258328.1
12
1557
XM_006722456.2
XP_006722519.1
13
1632
XM_011525991.1
XP_011524293.1
13
1632
XM_011525993.2
XP_011524295.1
13
1632
XM_011525994.2
XP_011524296.1
13
1632
XM_011525998.2
XP_011524300.1
12
1554
XM_011525999.1
XP_011524301.1
12
1557
XM_011526001.1
XP_011524303.1
11
1389
XM_011526002.1
XP_011524304.1
13
1632
XM_011526003.2
XP_011524305.1
13
1632
XM_011526006.1
XP_011524308.1
11
1389
XM_011526007.1
XP_011524309.1
12
1557
XM_017025751.1
XP_016881240.1
13
1632
XM_017025752.1
XP_016881241.1
12
1557
XM_017025753.1
XP_016881242.1
12
1557
XM_017025754.1
XP_016881243.1
13
1632
XM_017025755.1
XP_016881244.1
12
1557
XM_017025756.1
XP_016881245.1
13
1632
XM_017025757.1
XP_016881246.1
13
1632
XM_017025758.1
XP_016881247.1
13
1632
XM_017025759.1
XP_016881248.1
13
1632
XM_017025760.1
XP_016881249.1
12
1557
XM_017025761.1
XP_016881250.1
13
1632
XM_017025762.1
XP_016881251.1
12
1557
XM_017025763.1
XP_016881252.1
12
1554
XM_017025764.1
XP_016881253.1
13
1632
XM_017025765.1
XP_016881254.1
11
1389
XM_017025766.1
XP_016881255.1
12
1464
XM_017025767.1
XP_016881256.1
12
1464
XM_017025768.1
XP_016881257.1
11
1389
XM_017025769.1
XP_016881258.1
12
1557
XM_017025770.1
XP_016881259.1
11
1389
XM_017025771.1
XP_016881260.1
11
1389
XM_017025772.1
XP_016881261.1
11
1389
XM_017025773.1
XP_016881262.1
10
1320
XM_017025774.1
XP_016881263.1
10
1320
XM_017025775.1
XP_016881264.1
11
1389
XM_017025776.1
XP_016881265.1
11
1389
XM_017025777.1
XP_016881266.1
12
1557
GenBank mRNA
AB209765.1
-
8
907
AF078830.1
-
11
1373
AF078831.1
-
10
1304
AF078832.1
-
11
1205
AF078833.1
-
10
1205
AJ564845.1
-
12
1512
AK225889.1
-
12
1720
AK225948.1
-
12
1516
AK295401.1
-
13
1617
AK300759.1
-
11
1423
AK302004.1
-
13
1577
BC033242.1
-
11
1368
5432943
BC047353.1
-
11
1345
Y10746.1
-
11
1365
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
methylation-dependent chromatin silencing
transcription from RNA polymerase II promoter
heart development
aging
response to fluoxetine
neuron differentiation
response to nutrient levels
response to estradiol
response to cocaine
regulation of DNA methylation
negative regulation of transcription, DNA-templated
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
methyl-CpG binding
double-stranded methylated DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline