Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 3 | 933 | ||
NM_001286126.1 | NP_001273055.1 | 3 | 933 | |||
NM_001286128.1 | NP_001273057.1 | 3 | 933 | |||
NM_001286129.1 | NP_001273058.1 | 3 | 933 | |||
NM_018650.4 | NP_061120.3 | 3 | 933 | |||
XM_005273134.4 | XP_005273191.1 | 3 | 800 | |||
XM_006711326.3 | XP_006711389.1 | 2 | 55 | |||
XM_011509561.2 | XP_011507863.1 | 3 | 929 | |||
XM_011509562.2 | XP_011507864.1 | 3 | 518 | |||
XM_017001305.1 | XP_016856794.1 | 4 | 390 | |||
XM_017001306.1 | XP_016856795.1 | 3 | 402 | |||
XM_017001307.1 | XP_016856796.1 | 4 | 830 | |||
GenBank mRNA | AB040910.1 | - | 3 | 890 | ||
AF154845.1 | - | 3 | 559 | |||
AK098103.1 | - | 4 | 523 | |||
AK295506.1 | - | 3 | 431 | |||
AK314614.1 | - | 3 | 593 | |||
BC113869.1 | - | 3 | 469 | 40036731 | ||
BC114478.1 | - | 3 | 469 | 40036730 | ||
KJ534879.1 | - | 3 | 293 | |||
KJ534880.1 | - | 3 | 293 | |||
KJ534881.1 | - | 3 | 293 | |||
KJ535089.1 | - | 3 | 292 |