Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286124.1 | NP_001273053.1 | 18 | 4510 | ||
NM_001286126.1 | NP_001273055.1 | 17 | 4462 | |||
NM_001286128.1 | NP_001273057.1 | 16 | 4396 | |||
NM_018650.4 | NP_061120.3 | 18 | 4507 | |||
XM_005273134.4 | XP_005273191.1 | 17 | 4332 | |||
XM_006711326.3 | XP_006711389.1 | 18 | 3771 | |||
XM_011509561.2 | XP_011507863.1 | 18 | 4506 | |||
XM_011509562.2 | XP_011507864.1 | 18 | 4123 | |||
XM_017001305.1 | XP_016856794.1 | 19 | 3967 | |||
XM_017001306.1 | XP_016856795.1 | 18 | 4004 | |||
XM_017001307.1 | XP_016856796.1 | 18 | 4359 | |||
GenBank mRNA | AB040910.1 | - | 16 | 4353 | ||
AL512724.1 | - | 1 | 1854 |