Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000443.3 | NP_000434.1 | 17 | 2259 | ||
NM_018849.2 | NP_061337.1 | 17 | 2259 | |||
NM_018850.2 | NP_061338.1 | 17 | 2259 | |||
XM_011516308.2 | XP_011514610.2 | 17 | 2283 | |||
XM_011516309.2 | XP_011514611.2 | 17 | 2283 | |||
XM_011516310.2 | XP_011514612.2 | 17 | 2283 | |||
XM_011516311.2 | XP_011514613.2 | 17 | 2283 | |||
XM_011516312.2 | XP_011514614.2 | 17 | 2283 | |||
XM_011516313.2 | XP_011514615.1 | 17 | 2282 | |||
XM_011516315.2 | XP_011514617.2 | 12 | 1659 | |||
XM_017012323.1 | XP_016867812.1 | 17 | 2270 | |||
GenBank mRNA | BC042531.1 | - | 17 | 2295 | 4837724 | |
M23234.1 | - | 17 | 2215 |