Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319108.1 | NP_001306037.1 | 3 | 1758 | ||
NM_020530.5 | NP_065391.1 | 3 | 1577 | |||
GenBank mRNA | BC011589.1 | - | 3 | 1574 | 4548943 |