Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270523.1 | NP_001257452.1 | 5 | 1025 | ||
NM_001270524.1 | NP_001257453.1 | 4 | 1178 | |||
NM_001270525.1 | NP_001257454.1 | 3 | 972 | |||
NM_021728.3 | NP_068374.1 | 5 | 1049 | |||
NM_172337.2 | NP_758840.1 | 3 | 948 | |||
NR_073034.1 | 4 | 873 | ||||
NR_073036.1 | 2 | 796 | ||||
GenBank mRNA | AB593056.1 | - | 2 | 799 | ||
AB593057.1 | - | 3 | 718 | |||
AB593058.1 | - | 5 | 1049 | |||
AF093138.1 | - | 3 | 741 | |||
AK314271.1 | - | 4 | 1081 | |||
BC032579.1 | - | 5 | 1050 | 5493541 |