Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021963.3 | NP_068798.1 | 1 | 960 | ||
GenBank mRNA | AB027013.1 | - | 1 | 934 | ||
AF136178.1 | - | 1 | 908 | |||
AK291297.1 | - | 1 | 918 | |||
AK303676.1 | - | 2 | 553 | |||
AK316569.1 | - | 1 | 960 | |||
BC026325.1 | - | 1 | 882 | 4798964 | ||
CR457095.1 | - | 1 | 578 |