Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200001.1 | NP_001186930.1 | 9 | 1804 | ||
NM_024408.3 | NP_077719.2 | 9 | 1804 | |||
XM_005270901.3 | XP_005270958.1 | 10 | 4118 | |||
XM_011541519.2 | XP_011539821.1 | 9 | 6912 | |||
XM_011541520.2 | XP_011539822.1 | 9 | 3822 | |||
XM_017001372.1 | XP_016856861.1 | 9 | 1474 | |||
XM_017001373.1 | XP_016856862.1 | 9 | 1453 | |||
GenBank mRNA | AF308601.1 | - | 9 | 1519 | ||
AF315356.1 | - | 9 | 1519 | |||
AK307843.1 | - | 8 | 1645 | |||
BC071562.1 | - | 9 | 1753 | 4822955 | ||
GQ231534.1 | - | 9 | 1666 |