Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024408.3 | NP_077719.2 | 34 | 9292 | ||
XM_005270901.3 | XP_005270958.1 | 35 | 11606 | |||
XM_011541519.2 | XP_011539821.1 | 34 | 14400 | |||
XM_011541520.2 | XP_011539822.1 | 34 | 11310 | |||
XM_017001372.1 | XP_016856861.1 | 34 | 8962 | |||
XM_017001373.1 | XP_016856862.1 | 34 | 8941 | |||
GenBank mRNA | AF308601.1 | - | 34 | 9007 | ||
AF315356.1 | - | 34 | 9007 | |||
AK027869.1 | - | 8 | 4130 | |||
AL049386.1 | - | 1 | 432 | |||
AL133036.1 | - | 1 | 2969 |