Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190892.1 | NP_001177821.1 | 3 | 973 | ||
NM_001190898.2 | NP_001177827.1 | 4 | 1182 | |||
NM_001190899.2 | NP_001177828.1 | 3 | 1125 | |||
NM_001190900.1 | NP_001177829.1 | 4 | 872 | |||
NM_024411.4 | NP_077722.1 | 4 | 1185 | |||
XM_011529244.1 | XP_011527546.1 | 3 | 865 | |||
XM_011529245.1 | XP_011527547.1 | 3 | 1056 | |||
XM_011529246.2 | XP_011527548.1 | 3 | 1043 | |||
XM_011529247.1 | XP_011527549.1 | 3 | 852 | |||
XM_011529248.1 | XP_011527550.1 | 3 | 965 | |||
XM_011529249.2 | XP_011527551.1 | 3 | 1595 | |||
XM_011529250.2 | XP_011527552.1 | 3 | 1006 | |||
XM_017027878.1 | XP_016883367.1 | 3 | 855 | |||
GenBank mRNA | AK289618.1 | - | 3 | 973 | ||
AK295512.1 | - | 2 | 447 | |||
AK308747.1 | - | 2 | 797 | |||
AK316076.1 | - | 4 | 822 | |||
BC026334.1 | - | 4 | 958 | 4819025 |