Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033045.3 | NP_149034.2 | 4 | 917 | ||
XM_011538335.2 | XP_011536637.1 | 5 | 1032 | |||
GenBank mRNA | AJ628420.1 | - | 4 | 863 | ||
AK314030.1 | - | 4 | 936 | |||
BC069647.1 | - | 4 | 917 | 7262449 |