Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177969.1 | NP_001171440.1 | 6, 7 | 797 | ||
NM_001177970.1 | NP_001171441.1 | 6, 7 | 797 | |||
NM_001177971.1 | NP_001171442.1 | 6, 7 | 797 | |||
NM_053276.3 | NP_444506.2 | 6, 7 | 797 | |||
XM_017004325.1 | XP_016859814.1 | 6, 7 | 802 | |||
XM_017004327.1 | XP_016859816.1 | 4, 5 | 700 | |||
GenBank mRNA | AF063833.1 | - | 6, 7 | 621 | ||
AK056772.1 | - | 6, 7 | 716 | |||
AY358338.1 | - | 6, 7 | 682 | |||
BC128260.1 | - | 6, 7 | 610 | 40117956 |