Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322468.1 | NP_001309397.1 | 19765 | |||
NM_004532.5 | NP_004523.3 | 7 | 1039 | |||
NM_018406.6 | NP_060876.5 | 13747 | ||||
NM_138297.4 | NP_612154.2 | 6 | 886 | |||
GenBank mRNA | AJ010901.1 | - | 7 | 823 | ||
AJ242541.1 | - | 8 | 1048 | |||
AJ242543.1 | - | 7 | 747 | |||
AJ242544.1 | - | 7 | 719 | |||
AJ242546.1 | - | 7 | 823 | |||
AJ242547.1 | - | 7 | 823 | |||
AJ242548.1 | - | 7 | 823 | |||
AJ242549.1 | - | 7 | 923 | |||
AJ242550.1 | - | 6 | 770 | |||
AJ276359.1 | - | 9 | 3933 | |||
AJ277412.1 | - | 8 | 3777 | |||
AJ400633.1 | - | 9 | 3947 | |||
AJ400849.1 | - | 9 | 3857 | |||
AJ400850.1 | - | 9 | 3857 | |||
AJ400851.1 | - | 9 | 3947 | |||
AJ400852.1 | - | 8 | 3701 | |||
AJ400853.1 | - | 8 | 3673 | |||
AJ400854.1 | - | 8 | 3778 | |||
AJ400855.1 | - | 8 | 3749 | |||
AJ400856.1 | - | 9 | 3904 | |||
AJ400857.1 | - | 9 | 3933 | |||
AJ400858.1 | - | 9 | 3933 | |||
EF091824.1 | - | 10 | 3808 |