Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138733.4 | NP_620061.2 | 1 | 714 | ||
GenBank mRNA | AK312770.1 | - | 1 | 681 | ||
BC038843.1 | - | 1 | 671 | 5741907 | ||
FJ224296.1 | - | 1 | 631 |