Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 2 | 238 | ||
NM_001127717.2 | NP_001121189.2 | 39 | 5281 | |||
NM_001127718.2 | NP_001121190.2 | 2 | 238 | |||
NM_198129.2 | NP_937762.2 | 39 | 5281 | |||
XM_011525978.2 | XP_011524280.1 | 39 | 5308 | |||
XM_011525979.2 | XP_011524281.1 | 39 | 5299 | |||
XM_011525980.2 | XP_011524282.1 | 39 | 5290 | |||
XM_011525981.2 | XP_011524283.1 | 38 | 5176 | |||
XM_011525982.2 | XP_011524284.1 | 39 | 5308 | |||
XM_017025743.1 | XP_016881232.1 | 22 | 2992 | |||
XM_017025744.1 | XP_016881233.1 | 12 | 1673 | |||
GenBank mRNA | AB107369.1 | - | 39 | 5040 | ||
AY327114.1 | - | 2 | 213 | |||
AY327115.1 | - | 39 | 5040 | |||
AY327116.1 | - | 39 | 5040 | |||
DA862923.1 | - | 2 | 238 | |||
L34155.1 | - | 2 | 180 | |||
X84392.1 | - | 2 | 215 | |||
X84900.1 | - | 4 | 459 | |||
X85107.1 | - | 2 | 231 | |||
X85108.1 | - | 4 | 459 |