Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000227.4 | NP_000218.3 | 10 | 1186 | ||
NM_198129.2 | NP_937762.2 | 47 | 6229 | |||
XM_011525978.2 | XP_011524280.1 | 47 | 6256 | |||
XM_011525979.2 | XP_011524281.1 | 47 | 6247 | |||
XM_011525980.2 | XP_011524282.1 | 47 | 6238 | |||
XM_011525981.2 | XP_011524283.1 | 46 | 6124 | |||
XM_011525982.2 | XP_011524284.1 | 47 | 6256 | |||
XM_017025743.1 | XP_016881232.1 | 30 | 3940 | |||
XM_017025744.1 | XP_016881233.1 | 20 | 2621 | |||
GenBank mRNA | AB107369.1 | - | 47 | 5988 | ||
AB208853.1 | - | 2 | 214 | |||
AY327115.1 | - | 47 | 5988 | |||
BC043618.1 | - | 4 | 566 | |||
L34155.1 | - | 10 | 1128 | |||
X84900.1 | - | 12 | 1407 | |||
X85107.1 | - | 10 | 1179 | |||
X85108.1 | - | 12 | 1407 |