Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004255.3 | NP_004246.2 | 2, 3 | 365 | ||
GenBank mRNA | AI815421.1 | - | 2, 3 | 379 | ||
AK026615.1 | - | 2, 3 | 255 | |||
AK026623.1 | - | 2, 3 | 259 | |||
BC024240.2 | - | 2, 3 | 339 | 4815806 | ||
CR407649.1 | - | 2, 3 | 225 | |||
DQ987236.1 | - | 2, 3 | 225 | |||
DQ987237.1 | - | 2, 3 | 225 | |||
M22760.1 | - | 2, 3 | 244 |