Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002401.3 | NP_002392.2 | 16 | 4326 | ||
NM_203351.1 | NP_976226.1 | 17 | 4419 | |||
GenBank mRNA | BC010464.1 | - | 1 | 1654 | ||
BM800091.1 | - | 1 | 560 |
Gene Symbol: LIMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA251318.1 | - | 1 | 22 |